Assay Details
Target Gene Details
Entrez Gene ID: | 26524 |
Gene Name: | large tumor suppressor kinase 2 |
Gene Aliases: |
KPM |
Location: |
Chr.13:20973036-21061583 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LATS2 | NM_014572.2 | 8 | 3803 | NP_055387.2 |
| XM_005266342.1 | 8 | 3841 | XP_005266399.1 | |
| XM_011535042.2 | 7 | 3185 | XP_011533344.1 | |
| XM_017020541.1 | 8 | 3206 | XP_016876030.1 | |
| XM_017020542.1 | 7 | 3662 | XP_016876031.1 | |
| AB028019.1 | 7 | 3237 | BAA92381.1 | |
| AB209897.1 | 7 | 5414 | BAD93134.1 | |
| AF207547.1 | 8 | 3735 | AAF80561.1 | |
| AK123520.1 | 1 | 1696 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9059 | Chr.13:20963374 - 20996732 on Build GRCh38 | Gain |
|
| nsv1037082 | Chr.13:20932196 - 21204957 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map