Assay Details
Target Gene Details
Entrez Gene ID: | 133121 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 6 |
Gene Aliases: |
NPP6 |
Location: |
Chr.4:184088706-184217961 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENPP6 | NM_153343.3 | 8 | 3325 | NP_699174.1 |
| AK021943.1 | 1 | 1251 | ||
| BC035035.2 | 8 | 3230 | AAH35035.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521658 | Chr.4:184057338 - 184121636 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map