Assay Details
Target Gene Details
Entrez Gene ID: | 9938 |
Gene Name: | Rho GTPase activating protein 25 |
Gene Aliases: |
HEL-S-308, KAIA0053 |
Location: |
Chr.2:68710505-68826833 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP25 | NM_001007231.2 | 1 | 386 | NP_001007232.2 |
| NM_001166277.1 | 1 | 386 | NP_001159749.1 | |
| XM_005264675.2 | 1 | 93 | XP_005264732.1 | |
| XM_005264676.2 | 1 | 91 | XP_005264733.1 | |
| XM_011533207.2 | XP_011531509.1 | |||
| XM_011533208.2 | 1 | 58 | XP_011531510.1 | |
| AK290396.1 | 1 | 64 | ||
| AK297056.1 | 1 | 264 | ||
| AK303591.1 | ||||
| AK310742.1 | 1 | 91 | ||
| BX443252.2 | 1 | 331 | ||
| CD695161.1 | 1 | 409 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3862n100 | Chr.2:68727253 - 68771874 on Build GRCh38 | Gain |
|
| nsv953141 | Chr.2:68733769 - 68781868 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map