Assay Details
Target Gene Details
Entrez Gene ID: | 221830 |
Gene Name: | TWIST neighbor |
Gene Aliases: |
- |
Location: |
Chr.7:19695462-19709037 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TWISTNB | NM_001002926.1 | 4 | 3607 | NP_001002926.1 |
| AK090846.1 | 2 | 2998 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1018601 | Chr.7:19232737 - 19969258 on Build GRCh38 | Gain |
|
| nsv524807 | Chr.7:19674533 - 19764302 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map