Assay Details
Target Gene Details
Entrez Gene ID: | 2709 |
Gene Name: | gap junction protein beta 5 |
Gene Aliases: |
CX31.1 |
Location: |
Chr.1:34754777-34758513 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJB5 | NM_005268.3 | NP_005259.1 | ||
| XM_005270751.3 | XP_005270808.1 | |||
| AF052693.1 | AAD18005.1 | |||
| AK129509.1 | ||||
| BC004379.1 | AAH04379.1 |
More Information
Set Membership: |
|

Genomic Map