Assay Details
Target Gene Details
Entrez Gene ID: | 2110 |
Gene Name: | electron transfer flavoprotein dehydrogenase |
Gene Aliases: |
ETFQO, MADD |
Location: |
Chr.4:158672101-158708713 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ETFDH | NM_001281737.1 | 6 | 933 | NP_001268666.1 |
| NM_001281738.1 | 5 | 630 | NP_001268667.1 | |
| NM_004453.3 | 7 | 1074 | NP_004444.2 | |
| AK293737.1 | 5 | 615 | ||
| AK304838.1 | 6 | 695 | ||
| AK314803.1 | 7 | 1051 | ||
| BC011890.1 | 7 | 876 | AAH11890.1 | |
| BX538129.1 | 8 | 746 | CAD98030.1 | |
| EU093082.1 | 7 | 770 | ||
| EU093083.1 | ||||
| S69232.1 | 7 | 838 | AAC60628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830125 | Chr.4:158691163 - 158848360 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map