Assay Details
Target Gene Details
Entrez Gene ID: | 378708 |
Gene Name: | apoptosis-inducing, TAF9-like domain 1 |
Gene Aliases: |
CENP-S, CENPS, FAAP16, MHF1 |
Location: |
Chr.1:10430102-10442815 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APITD1 | NM_199294.2 | NP_954988.1 | ||
| NR_036462.1 | ||||
| BC029430.1 | AAH29430.1 | |||
| BU902753.1 | ||||
| CA311915.1 |
Target Gene Details
Entrez Gene ID: | 100526739 |
Gene Name: | APITD1-CORT readthrough |
Gene Aliases: |
- |
Location: |
Chr.1:10430102-10452003 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APITD1-CORT | NM_001243768.1 | NP_001230697.1 | ||
| NM_001270517.1 | NP_001257446.1 | |||
| NM_198544.3 | NP_940946.1 | |||
| NM_199006.2 | NP_950171.2 | |||
| NR_037187.1 | ||||
| AA858169.1 | ||||
| AF516753.1 | AAM61954.1 | |||
| AF521016.1 | AAM70482.1 | |||
| CF127457.1 | ||||
| CF128105.1 | ||||
| HY054077.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2764192 | Chr.1:10323676 - 10534129 on Build GRCh38 | Gain |
|
| esv2758920 | Chr.1:10178012 - 10522497 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map