Assay Details
Target Gene Details
Entrez Gene ID: | 114879 |
Gene Name: | oxysterol binding protein like 5 |
Gene Aliases: |
OBPH1, ORP5 |
Location: |
Chr.11:3087116-3165352 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OSBPL5 | NM_001144063.1 | 7 | 706 | NP_001137535.1 |
| NM_020896.3 | 8 | 910 | NP_065947.1 | |
| NM_145638.2 | 7 | 699 | NP_663613.1 | |
| XM_011519873.2 | 8 | 1249 | XP_011518175.1 | |
| XM_017017163.1 | 8 | 1257 | XP_016872652.1 | |
| XM_017017164.1 | 3 | 828 | XP_016872653.1 | |
| AB074006.1 | 8 | 851 | BAB85686.1 | |
| AF392453.1 | 8 | 823 | AAL40666.1 | |
| AF410430.1 | 7 | 768 | AAK98617.1 | |
| AK055617.1 | 3 | 244 | ||
| AK056510.1 | 8 | 868 | ||
| AK293000.1 | 7 | 655 | ||
| AK297475.1 | ||||
| AK301003.1 | 6 | 546 | ||
| AK303205.1 | 8 | 728 | ||
| AK309411.1 | 8 | 888 | ||
| AL136918.1 | 8 | 823 | CAB66852.1 | |
| BC032646.1 | 7 | 620 | AAH32646.2 | |
| BC039579.1 | 8 | 821 | AAH39579.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951293 | Chr.11:3089571 - 3111670 on Build GRCh38 | Deletion |
|
| nsv8766 | Chr.11:2971449 - 3205956 on Build GRCh38 | Loss |
|
| nsv509384 | Chr.11:3088735 - 3189373 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map