Assay Details
Target Gene Details
Entrez Gene ID: | 64800 |
Gene Name: | EF-hand calcium binding domain 6 |
Gene Aliases: |
DJBP, HSCBCIP1, dJ185D5.1 |
Location: |
Chr.22:43528549-43812403 on Build GRCh38 |
Assay Gene Location: | Within Exon 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB6 | NM_022785.3 | 32 | 4851 | NP_073622.2 |
| NM_198856.2 | 30 | 4493 | NP_942153.1 | |
| XM_005261704.2 | 32 | 5049 | XP_005261761.1 | |
| XM_005261705.3 | 31 | 4963 | XP_005261762.1 | |
| XM_011530316.1 | 33 | 5336 | XP_011528618.1 | |
| XM_011530317.2 | 31 | 4896 | XP_011528619.1 | |
| XM_011530318.2 | 31 | 4992 | XP_011528620.1 | |
| XM_011530319.2 | 30 | 4684 | XP_011528621.1 | |
| XM_011530320.2 | 33 | 4886 | XP_011528622.1 | |
| XM_011530321.2 | 30 | 4679 | XP_011528623.1 | |
| XM_011530322.2 | 31 | 4807 | XP_011528624.1 | |
| XM_011530323.2 | 28 | 4465 | XP_011528625.1 | |
| XM_011530325.2 | 26 | 4210 | XP_011528627.1 | |
| XM_011530327.2 | 23 | 3957 | XP_011528629.1 | |
| XM_011530329.1 | 19 | 3429 | XP_011528631.1 | |
| XM_017028910.1 | 33 | 5148 | XP_016884399.1 | |
| XM_017028911.1 | 33 | 4890 | XP_016884400.1 | |
| XM_017028912.1 | 29 | 4807 | XP_016884401.1 | |
| AA778573.1 | 1 | 83 | ||
| AB073862.1 | BAB71780.1 | |||
| AK027241.1 | BAB15703.1 | |||
| AK225596.1 | ||||
| AK225605.1 | ||||
| AK292411.1 | 32 | 4791 | ||
| AK292498.1 | 30 | 4470 | ||
| AL355841.1 | 7 | 1174 | CAB91065.1 | |
| AW139921.1 | 1 | 66 | ||
| BC039315.1 | 30 | 4346 | AAH39315.1 |
Target Gene Details
Entrez Gene ID: | 100874197 |
Gene Name: | EFCAB6 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.22:43516254-43536605 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB6-AS1 | NR_046563.1 | |||
| AK090843.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058659 | Chr.22:43522577 - 43700374 on Build GRCh38 | Loss |
|
| nsv589098 | Chr.22:43463583 - 43718462 on Build GRCh38 | Gain |
|
| nsv471205 | Chr.22:43467417 - 43718981 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map