Assay Details
Target Gene Details
Entrez Gene ID: | 91875 |
Gene Name: | tetratricopeptide repeat domain 5 |
Gene Aliases: |
Strap |
Location: |
Chr.14:20289131-20305994 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC5 | NM_138376.2 | 10 | 1702 | NP_612385.2 |
| AK074553.1 | 10 | 1685 | BAC11056.1 | |
| BC008647.1 | 10 | 1643 | AAH08647.1 | |
| BC030822.1 | 10 | 1646 | AAH30822.1 | |
| BC053538.1 | 10 | 1650 | AAH53538.1 | |
| BM998414.1 | 1 | 180 | ||
| BX640732.1 | 10 | 1581 | CAE45848.1 | |
| BX648584.1 | 9 | 4836 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3633679 | Chr.14:20210486 - 20289540 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map