Assay Details
Target Gene Details
Entrez Gene ID: | 6548 |
Gene Name: | solute carrier family 9 member A1 |
Gene Aliases: |
APNH, LIKNS, NHE-1, NHE1, PPP1R143 |
Location: |
Chr.1:27098809-27155130 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9A1 | NM_003047.4 | 4 | 1925 | NP_003038.2 |
| NR_046474.1 | 3 | 1464 | ||
| XM_011542021.2 | 5 | 1003 | XP_011540323.1 | |
| AF141350.1 | 4 | 1172 | AAF21350.1 | |
| AF141351.1 | 4 | 1172 | AAF21351.1 | |
| AF141352.1 | 4 | 1172 | AAF21352.1 | |
| AF141353.1 | 4 | 1172 | AAF21353.1 | |
| AF141354.1 | 4 | 1172 | AAF21354.1 | |
| AF141355.1 | 4 | 1172 | AAF21355.1 | |
| AF141356.1 | 4 | 1172 | AAF21356.1 | |
| AF141357.1 | 4 | 1172 | AAF21357.1 | |
| AF141358.1 | 4 | 1172 | AAF21358.1 | |
| AF141359.1 | 4 | 1172 | AAF21359.1 | |
| AF146430.1 | 4 | 1172 | AAF25592.1 | |
| AF146431.1 | 4 | 1172 | AAF25593.1 | |
| AF146432.1 | 4 | 1172 | AAF25594.1 | |
| AF146433.1 | 4 | 1172 | AAF25595.1 | |
| AF146434.1 | 4 | 1172 | AAF25596.1 | |
| AF146435.1 | 4 | 1172 | AAF25597.1 | |
| AF146436.1 | 4 | 1172 | AAF25598.1 | |
| AF146437.1 | 4 | 1172 | AAF25599.1 | |
| AF146438.1 | 4 | 1172 | AAF25600.1 | |
| AF146439.1 | 4 | 1172 | AAF25601.1 | |
| AK300432.1 | 3 | 816 | ||
| AK314190.1 | 4 | 1320 | ||
| BC012121.1 | 4 | 1393 | AAH12121.1 | |
| M81768.1 | 4 | 1182 | AAB59460.1 | |
| M96066.1 | 4 | 1706 | ||
| S68616.1 | 4 | 1706 | AAC60606.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834535 | Chr.1:27006585 - 27179926 on Build GRCh38 | Gain+Loss |
|
| nsv1000012 | Chr.1:26714969 - 27141528 on Build GRCh38 | Gain |
|
| nsv509058 | Chr.1:27068848 - 27109365 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map