Assay Details
Target Gene Details
Entrez Gene ID: | 4800 |
Gene Name: | nuclear transcription factor Y subunit alpha |
Gene Aliases: |
CBF-A, CBF-B, HAP2, NF-YA |
Location: |
Chr.6:41072968-41102407 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFYA | NM_002505.4 | 9 | 1117 | NP_002496.1 |
| NM_021705.3 | 8 | 1030 | NP_068351.1 | |
| BC039244.1 | 8 | 1283 | AAH39244.1 | |
| BM561405.1 | ||||
| GQ167198.1 | 8 | 936 | ||
| KU159426.1 | 8 | 1074 | ||
| M59079.1 | 6 | 702 | AAA35950.1 | |
| X59711.1 | 9 | 1113 | CAA42231.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv602970 | Chr.6:41070039 - 41258521 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map