Assay Details
Target Gene Details
Entrez Gene ID: | 55740 |
Gene Name: | enabled homolog (Drosophila) |
Gene Aliases: |
ENA, MENA, NDPP1 |
Location: |
Chr.1:225486832-225660045 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENAH | NM_001008493.1 | 15 | 11067 | NP_001008493.1 |
| NM_018212.4 | 14 | 11004 | NP_060682.2 | |
| XM_005273187.2 | 16 | 11107 | XP_005273244.1 | |
| XM_017001746.1 | 17 | 12342 | XP_016857235.1 | |
| XM_017001751.1 | 14 | 11352 | XP_016857240.1 | |
| XM_017001753.1 | 18 | 11146 | XP_016857242.1 | |
| XM_017001754.1 | 17 | 11090 | XP_016857243.1 | |
| XM_017001755.1 | 17 | 10833 | XP_016857244.1 | |
| XM_017001756.1 | 15 | 10764 | XP_016857245.1 | |
| XM_017001757.1 | 15 | 10667 | XP_016857246.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004851 | Chr.1:225426438 - 225496482 on Build GRCh38 | Gain |
|
| nsv433350 | Chr.1:225477582 - 225498323 on Build GRCh38 | Gain |
|
| nsv826842 | Chr.1:225470360 - 225498884 on Build GRCh38 | Gain |
|
| esv3588992 | Chr.1:225469902 - 225499153 on Build GRCh38 | Gain |
|
| esv3575750 | Chr.1:225471464 - 225496482 on Build GRCh38 | Gain |
|
| nsv832714 | Chr.1:225404631 - 225617694 on Build GRCh38 | Loss |
|
| nsv4720 | Chr.1:225485516 - 225530307 on Build GRCh38 | Deletion |
|
| nsv8880 | Chr.1:225468706 - 225500008 on Build GRCh38 | Gain |
|
| esv2759003 | Chr.1:225477992 - 225515783 on Build GRCh38 | Gain |
|
| nsv441740 | Chr.1:225471515 - 225496494 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map