Assay Details
Target Gene Details
Entrez Gene ID: | 29122 |
Gene Name: | protease, serine 50 |
Gene Aliases: |
CT20, TSP50 |
Location: |
Chr.3:46712116-46717883 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS50 | NM_013270.4 | NP_037402.1 | ||
| BC033016.1 | AAH33016.1 | |||
| BC037775.2 | AAH37775.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516623 | Chr.3:46638573 - 46811189 on Build GRCh38 | Loss |
|
| esv3596066 | Chr.3:46626410 - 46715170 on Build GRCh38 | Gain |
|
| nsv954478 | Chr.3:46657411 - 46717710 on Build GRCh38 | Deletion |
|
| nsv590179 | Chr.3:46687169 - 46748907 on Build GRCh38 | Loss |
|
| nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73831200] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map