Assay Details
Target Gene Details
Entrez Gene ID: | 2647 |
Gene Name: | biogenesis of lysosomal organelles complex 1 subunit 1 |
Gene Aliases: |
BLOS1, BORCS1, GCN5L1, MICoA, RT14 |
Location: |
Chr.12:55716034-55719707 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLOC1S1 | NM_001487.3 | NP_001478.2 | ||
| NR_037655.1 | ||||
| NR_037656.1 | ||||
| NR_037657.1 | ||||
| AI673441.1 | ||||
| AI816035.1 | ||||
| AK307069.1 | 3 | 674 | ||
| BC130640.1 | ||||
| BC132795.1 | ||||
| BQ899204.1 | ||||
| BU601406.1 | ||||
| CN263524.1 | ||||
| D64007.1 | BAA10887.1 | |||
| DA243440.1 | ||||
| DA864590.1 | ||||
| S82447.1 | AAB37682.1 |
Target Gene Details
Entrez Gene ID: | 100528022 |
Gene Name: | BLOC1S1-RDH5 readthrough |
Gene Aliases: |
- |
Location: |
Chr.12:55716034-55724742 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLOC1S1-RDH5 | NR_037658.1 | |||
| AK096656.1 | ||||
| AK307141.1 | ||||
| BG722558.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832423 | Chr.12:55687038 - 55859914 on Build GRCh38 | Gain |
|
| nsv559004 | Chr.12:55704254 - 55777109 on Build GRCh38 | Loss |
|
| nsv525231 | Chr.12:55683135 - 55730548 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map