Assay Details
Target Gene Details
Entrez Gene ID: | 219285 |
Gene Name: | sterile alpha motif domain containing 9 like |
Gene Aliases: |
C7orf6, DRIF2, UEF1 |
Location: |
Chr.7:93130054-93148401 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SAMD9L | NM_001303496.1 | 5 | 5920 | NP_001290425.1 |
NM_001303497.1 | 6 | 5749 | NP_001290426.1 | |
NM_001303498.1 | 5 | 5485 | NP_001290427.1 | |
NM_001303500.1 | 4 | 5353 | NP_001290429.1 | |
NM_152703.3 | 5 | 6125 | NP_689916.2 | |
XM_005250193.3 | 6 | 5557 | XP_005250250.1 | |
XM_006715890.1 | 2 | 4926 | XP_006715953.1 | |
XM_011515903.2 | 6 | 5810 | XP_011514205.1 | |
XM_017011822.1 | 4 | 5874 | XP_016867311.1 | |
XM_017011823.1 | 5 | 5630 | XP_016867312.1 | |
AB095926.1 | 5 | 6093 | ||
AF474973.1 | 6 | 5502 | AAQ05771.1 | |
AK026672.1 | 1 | 381 | ||
AK097204.1 | 1 | 2307 | BAC04975.1 | |
AK304779.1 | 1 | 3894 | ||
AL832264.1 | 1 | 2991 | ||
AL834232.1 | 1 | 1633 | CAD38910.1 | |
AY195587.1 | 3 | 450 | ||
BC127117.1 | 5 | 5361 | ||
BC127118.1 | 4 | 5737 |

More Information
Additional Information:
For this assay, SNP(s) [rs116965992] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
![]() ![]() ![]() |
