Assay Details
Target Gene Details
Entrez Gene ID: | 182 |
Gene Name: | jagged 1 |
Gene Aliases: |
AGS, AHD, AWS, CD339, HJ1, JAGL1 |
Location: |
Chr.20:10637684-10674046 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAG1 | NM_000214.2 | NP_000205.1 | ||
| XM_017027707.1 | XP_016883196.1 | |||
| AF003837.1 | AAC51731.1 | |||
| AF028593.1 | AAB84053.1 | |||
| AK302554.1 | ||||
| AK312722.1 | ||||
| BC098393.1 | AAH98393.1 | |||
| BC126205.1 | ||||
| BC126207.1 | ||||
| U61276.1 | AAB39007.1 | |||
| U73936.1 | AAC52020.1 | |||
| U77720.1 | AAC51323.1 |
Target Gene Details
Entrez Gene ID: | 102465525 |
Gene Name: | microRNA 6870 |
Gene Aliases: |
hsa-mir-6870 |
Location: |
Chr.20:10649636-10649695 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR6870 | NR_106930.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3291 | Chr.20:10623650 - 10655782 on Build GRCh38 | Insertion |
|
| nsv522874 | Chr.20:9918791 - 10838562 on Build GRCh38 | Loss |
|
| nsv833920 | Chr.20:10460192 - 10651051 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map