Assay Details
Target Gene Details
Entrez Gene ID: | 9447 |
Gene Name: | absent in melanoma 2 |
Gene Aliases: |
PYHIN4 |
Location: |
Chr.1:159059226-159132351 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AIM2 | NM_004833.1 | 5 | 1147 | NP_004824.1 |
| XM_005245616.4 | 4 | 918 | XP_005245673.1 | |
| XM_017002848.1 | 6 | 2374 | XP_016858337.1 | |
| AF024714.1 | 5 | 1147 | AAB81613.1 | |
| AK292042.1 | 5 | 1056 | ||
| BC010940.1 | 5 | 1149 | AAH10940.1 |
More Information
Set Membership: |
|

Genomic Map