Assay Details
Target Gene Details
Entrez Gene ID: | 726 |
Gene Name: | calpain 5 |
Gene Aliases: |
ADNIV, HTRA3, VRNI, nCL-3 |
Location: |
Chr.11:77066945-77126155 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPN5 | NM_004055.4 | 2 | 239 | NP_004046.2 |
| XM_011545225.1 | 3 | 360 | XP_011543527.1 | |
| XM_017018223.1 | 2 | 162 | XP_016873712.1 | |
| AB209087.1 | 2 | 168 | BAD92324.1 | |
| AK094150.1 | 2 | 191 | ||
| AK128124.1 | 3 | 360 | BAC87282.1 | |
| AK310978.1 | 3 | 280 | ||
| AK313132.1 | 2 | 213 | ||
| BC018123.1 | 2 | 239 | AAH18123.1 | |
| U94346.1 | 2 | 200 | AAC51869.1 | |
| Y10552.1 | 1 | 97 | CAA71584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1040996 | Chr.11:77037138 - 77121673 on Build GRCh38 | Gain |
|
| nsv825998 | Chr.11:77045911 - 77178724 on Build GRCh38 | Gain |
|
| dgv1230n100 | Chr.11:77064328 - 77184255 on Build GRCh38 | Gain |
|
| nsv1044491 | Chr.11:77069929 - 77126506 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map