Assay Details
Target Gene Details
Entrez Gene ID: | 84950 |
Gene Name: | pre-mRNA processing factor 38A |
Gene Aliases: |
Prp38 |
Location: |
Chr.1:52404547-52419723 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF38A | NM_032864.3 | 10 | 2212 | NP_116253.2 |
| XM_011542315.2 | 10 | 2233 | XP_011540617.1 | |
| AK027842.1 | 9 | 1786 | BAB55405.1 | |
| AK092038.1 | 10 | 2195 | BAC03796.1 | |
| BE465896.1 |
More Information
Set Membership: |
|

Genomic Map