Assay Details
Target Gene Details
Entrez Gene ID: | 7965 |
Gene Name: | aminoacyl tRNA synthetase complex interacting multifunctional protein 2 |
Gene Aliases: |
JTV-1, JTV1, P38 |
Location: |
Chr.7:6009241-6023834 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AIMP2 | NM_001326606.1 | 4 | 1002 | NP_001313535.1 |
| NM_001326607.1 | 3 | 915 | NP_001313536.1 | |
| NM_001326609.1 | 5 | 1208 | NP_001313538.1 | |
| NM_001326611.1 | 4 | 921 | NP_001313540.1 | |
| NM_006303.3 | 4 | 1122 | NP_006294.2 | |
| XM_005249847.3 | 4 | 1336 | XP_005249904.1 | |
| XM_005249848.2 | 4 | 1012 | XP_005249905.1 | |
| XM_005249850.3 | 4 | 933 | XP_005249907.1 | |
| XM_006715772.2 | 4 | 907 | XP_006715835.1 | |
| XM_011515519.1 | 4 | 937 | XP_011513821.1 | |
| AK313037.1 | ||||
| BC002853.1 | 4 | 1082 | AAH02853.1 | |
| BC010156.1 | 4 | 1082 | AAH10156.1 | |
| BC013630.1 | 4 | 1090 | AAH13630.1 | |
| U24169.1 | 4 | 1122 | AAC50391.1 |
Target Gene Details
Entrez Gene ID: | 27102 |
Gene Name: | eukaryotic translation initiation factor 2 alpha kinase 1 |
Gene Aliases: |
HCR, HRI |
Location: |
Chr.7:6022247-6059229 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2AK1 | NM_001134335.1 | 15 | 2972 | NP_001127807.1 |
| NM_014413.3 | 15 | 2975 | NP_055228.2 | |
| AB037790.1 | 15 | 2900 | ||
| AF147094.1 | AAF66736.1 | |||
| AF255050.1 | AAF70289.1 | |||
| AK295712.1 | 15 | 2182 | ||
| AL136563.1 | CAB66498.1 | |||
| AL834494.1 | CAD39152.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034916 | Chr.7:5895149 - 6119339 on Build GRCh38 | Gain |
|
| nsv436904 | Chr.7:5035574 - 6713282 on Build GRCh38 | Insertion |
|
| nsv830894 | Chr.7:5850027 - 6032584 on Build GRCh38 | Loss |
|
| nsv605998 | Chr.7:5995434 - 6062403 on Build GRCh38 | Loss |
|
| nsv1018388 | Chr.7:5999082 - 6091538 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map