Assay Details
Target Gene Details
Entrez Gene ID: | 6734 |
Gene Name: | SRP receptor alpha subunit |
Gene Aliases: |
DP, SRPR, Sralpha |
Location: |
Chr.11:126255151-126268982 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRPRA | NM_001177842.1 | NP_001171313.1 | ||
| NM_003139.3 | NP_003130.2 | |||
| XM_017018179.1 | XP_016873668.1 | |||
| AK130382.1 | ||||
| AK303379.1 | ||||
| AK312377.1 | ||||
| BC001162.1 | AAH01162.1 | |||
| BC009110.1 | AAH09110.1 | |||
| BC013583.1 | AAH13583.1 | |||
| CR457025.1 | CAG33306.1 | |||
| X06272.1 | CAA29608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556530 | Chr.11:126264724 - 126393990 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map