Assay Details
Target Gene Details
Entrez Gene ID: | 5137 |
Gene Name: | phosphodiesterase 1C |
Gene Aliases: |
Hcam3, cam-PDE 1C, hCam-3 |
Location: |
Chr.7:31616777-32428224 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE1C |
Target Gene Details
Entrez Gene ID: | 10842 |
Gene Name: | protein phosphatase 1 regulatory subunit 17 |
Gene Aliases: |
C7orf16, GSBS |
Location: |
Chr.7:31687017-31709931 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R17 | NM_001145123.2 | 4 | 1629 | NP_001138595.1 |
| NM_006658.4 | 5 | 1782 | NP_006649.2 | |
| XM_011515094.1 | 6 | 1414 | XP_011513396.1 | |
| AF071789.1 | 4 | 1507 | AAD12588.1 | |
| AF097730.1 | 4 | 1504 | AAD13030.1 | |
| BC028094.1 | 5 | 1566 | AAH28094.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3612723 | Chr.7:31646533 - 31888101 on Build GRCh38 | Gain |
|
| esv3612724 | Chr.7:31646828 - 31721794 on Build GRCh38 | Gain |
|
| nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113797486] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map