Assay Details
Target Gene Details
Entrez Gene ID: | 10411 |
Gene Name: | Rap guanine nucleotide exchange factor 3 |
Gene Aliases: |
CAMP-GEFI, EPAC, EPAC1, HSU79275, bcm910 |
Location: |
Chr.12:47734670-47759106 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAPGEF3 | NM_001098531.2 | 28 | 3425 | NP_001092001.1 |
| NM_001098532.2 | 27 | 3189 | NP_001092002.1 | |
| NM_006105.5 | 28 | 2959 | NP_006096.2 | |
| XM_005268571.3 | 27 | 2935 | XP_005268628.1 | |
| XM_011537752.1 | 28 | 3442 | XP_011536054.1 | |
| XM_011537755.1 | 28 | 3004 | XP_011536057.1 | |
| XM_011537758.2 | 28 | 3054 | XP_011536060.1 | |
| AK055502.1 | 10 | 1740 | ||
| AK074444.1 | 12 | 2117 | ||
| AK092448.1 | 28 | 3397 | ||
| AK125545.1 | 24 | 2442 | ||
| AK290230.1 | 29 | 3090 | ||
| BC017728.1 | 28 | 3039 | ||
| BC092404.1 | 28 | 3028 | ||
| BQ007070.1 | 1 | 237 | ||
| U78168.1 | 28 | 2964 | AAD12740.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv696 | Chr.12:47720180 - 47754753 on Build GRCh38 | Insertion |
|
| nsv470289 | Chr.12:47716768 - 47786725 on Build GRCh38 | Gain |
|
| dgv94n21 | Chr.12:47725779 - 47821397 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map