Assay Details
Target Gene Details
Entrez Gene ID: | 167127 |
Gene Name: | UDP glycosyltransferase family 3 member A2 |
Gene Aliases: |
- |
Location: |
Chr.5:36035017-36071358 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT3A2 | NM_001168316.1 | 6 | 1682 | NP_001161788.1 |
| NM_174914.3 | 7 | 1784 | NP_777574.2 | |
| NR_031764.1 | 6 | 1252 | ||
| XM_011513988.2 | 8 | 1826 | XP_011512290.1 | |
| XM_017009150.1 | 7 | 1835 | XP_016864639.1 | |
| AK075383.1 | 7 | 1745 | BAC11583.1 | |
| AK300751.1 | 6 | 1643 | ||
| AK303154.1 | 6 | 1213 | ||
| AY358416.1 | 7 | 1719 | AAQ88782.1 | |
| BC103924.1 | 7 | 1733 | AAI03925.1 | |
| BC103925.1 | 7 | 1733 | AAI03926.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3604707 | Chr.5:35513484 - 36501574 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76748392] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map