Assay Details
Target Gene Details
Entrez Gene ID: | 509 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1 |
Gene Aliases: |
ATP5C, ATP5CL1 |
Location: |
Chr.10:7788104-7807801 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5C1 | NM_001001973.2 | NP_001001973.1 | ||
| NM_001320886.1 | NP_001307815.1 | |||
| NM_005174.3 | NP_005165.1 | |||
| XM_017016290.1 | XP_016871779.1 | |||
| AK292896.1 | ||||
| AK296803.1 | ||||
| BC000470.2 | AAH00470.1 | |||
| BC000931.3 | AAH00931.1 | |||
| BC013394.2 | AAH13394.1 | |||
| BC016812.1 | AAH16812.1 | |||
| BC020824.1 | AAH20824.1 | |||
| BC026049.1 | AAH26049.1 | |||
| BI603806.1 | ||||
| BQ893894.1 | ||||
| CR457403.1 | CAG33684.1 | |||
| D16562.1 | BAA03996.1 | |||
| D16563.1 | BAA03997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549933 | Chr.10:7586456 - 8160993 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map