Assay Details
Target Gene Details
Entrez Gene ID: | 168002 |
Gene Name: | dishevelled binding antagonist of beta catenin 2 |
Gene Aliases: |
C6orf116, DAPPER2, DPR2, bA503C24.7 |
Location: |
Chr.6:168292830-168319754 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DACT2 | NM_001286350.1 | 3 | 1131 | NP_001273279.1 |
| NM_001286351.1 | NP_001273280.1 | |||
| NM_214462.4 | 4 | 1654 | NP_999627.2 | |
| NR_104425.1 | 2 | 663 | ||
| XM_011535507.2 | 4 | 1416 | XP_011533809.1 | |
| AF318336.1 | 4 | 1623 | AAL55843.1 | |
| BC092498.1 | AAH92498.1 | |||
| BC111764.1 | 2 | 619 | AAI11765.1 | |
| BC111790.1 | 3 | 1087 | AAI11791.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830867 | Chr.6:168203477 - 168373698 on Build GRCh38 | Loss |
|
| esv2759493 | Chr.6:167836188 - 168314704 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79931308] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map