Assay Details
Target Gene Details
Entrez Gene ID: | 8995 |
Gene Name: | tumor necrosis factor superfamily member 18 |
Gene Aliases: |
AITRL, GITRL, TL6, TNLG2A, hGITRL |
Location: |
Chr.1:173041220-173050963 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFSF18 | NM_005092.3 | 3 | 625 | NP_005083.2 |
| AF117713.1 | AAD19695.1 | |||
| AF125303.1 | AAD22634.1 | |||
| AK313273.1 | ||||
| AY358868.1 | 3 | 578 | AAQ89227.1 | |
| BC069111.1 | AAH69111.1 | |||
| BC069319.1 | AAH69319.1 | |||
| BC093986.1 | 3 | 625 | AAH93986.1 | |
| BC112032.1 | 3 | 625 | AAI12033.1 | |
| BX473859.1 | 3 | 602 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831904 | Chr.1:172942970 - 173101289 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map