Assay Details
Target Gene Details
Entrez Gene ID: | 84153 |
Gene Name: | ribonuclease H2 subunit C |
Gene Aliases: |
AGS3, AYP1 |
Location: |
Chr.11:65717673-65720938 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASEH2C | NM_032193.3 | 1 | 193 | NP_115569.2 |
| AF312034.1 | AAL87739.1 | |||
| AF346606.1 | 1 | 193 | AAO49177.1 | |
| AK024627.1 | BAB14937.1 | |||
| AK225131.1 | ||||
| AK297660.1 | ||||
| BC023588.2 | 1 | 73 | AAH23588.1 | |
| BM921525.1 |
More Information
Set Membership: |
|

Genomic Map