Assay Details
Target Gene Details
Entrez Gene ID: | 196410 |
Gene Name: | methyltransferase like 7B |
Gene Aliases: |
ALDI |
Location: |
Chr.12:55681546-55684611 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL7B | NM_152637.2 | 2 | 1245 | NP_689850.2 |
| AY358508.1 | 2 | 1113 | AAQ88872.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525231 | Chr.12:55683135 - 55730548 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map