Assay Details
Target Gene Details
Entrez Gene ID: | 11126 |
Gene Name: | CD160 molecule |
Gene Aliases: |
BY55, NK1, NK28 |
Location: |
Chr.1:145719312-145739288 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CD160 | NM_007053.3 | NP_008984.1 | ||
NR_103845.1 | ||||
XM_005272929.3 | XP_005272986.1 | |||
XM_011509104.2 | XP_011507406.1 | |||
AF060981.1 | AAC72302.1 | |||
AK128370.1 | ||||
BC014465.1 | AAH14465.1 | |||
CD638675.1 | ||||
CR541867.1 | CAG46665.1 | |||
CR541888.1 | CAG46686.1 | |||
EU016100.1 | ||||
EU016101.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3587421 | Chr.1:145649153 - 145751059 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
dgv331n100 | Chr.1:145642988 - 145827134 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv3572342 | Chr.1:145601946 - 145809476 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv469752 | Chr.1:145639438 - 145790042 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv831392 | Chr.1:145632374 - 145813165 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
esv3894089 | Chr.1:145672100 - 145824903 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv547670 | Chr.1:145689573 - 145778504 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv532n54 | Chr.1:145624561 - 145813526 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
dgv33n27 | Chr.1:145687602 - 145824903 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
dgv95e212 | Chr.1:145631218 - 145812561 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
