Assay Details
Target Gene Details
Entrez Gene ID: | 56265 |
Gene Name: | carboxypeptidase X (M14 family), member 1 |
Gene Aliases: |
CPX1, CPXM |
Location: |
Chr.20:2794069-2800646 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPXM1 | NM_001184699.1 | NP_001171628.1 | ||
| NM_019609.4 | 11 | 1712 | NP_062555.1 | |
| AK027661.1 | 11 | 1648 | BAB55275.1 | |
| AK075508.1 | 7 | 924 | BAC11661.1 | |
| AK075527.1 | BAC11672.1 | |||
| AK303142.1 | 8 | 1191 | ||
| AK315027.1 | ||||
| AY358956.1 | 11 | 1668 | AAQ89315.1 | |
| BC063430.1 | 11 | 1689 | AAH63430.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952977 | Chr.20:2795055 - 2800754 on Build GRCh38 | Deletion |
|
| nsv1057464 | Chr.20:2650623 - 2989686 on Build GRCh38 | Gain |
|
| nsv510780 | Chr.20:2746049 - 2832153 on Build GRCh38 | Deletion |
|
| nsv522009 | Chr.20:2745032 - 3030542 on Build GRCh38 | Gain |
|
| nsv833895 | Chr.20:2730164 - 2882236 on Build GRCh38 | Loss |
|
| esv2762056 | Chr.20:2740745 - 2944353 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map