Assay Details
Target Gene Details
Entrez Gene ID: | 6923 |
Gene Name: | transcription elongation factor B subunit 2 |
Gene Aliases: |
ELOB, SIII |
Location: |
Chr.16:2771414-2777296 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCEB2 | NM_007108.3 | 4 | 735 | NP_009039.1 |
| NM_207013.2 | NP_996896.1 | |||
| AI191983.1 | ||||
| BC013306.2 | 3 | 689 | AAH13306.1 | |
| BM553320.1 | 4 | 750 | ||
| BM700019.1 | ||||
| BM921799.1 | 3 | 703 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760067 | Chr.16:2486824 - 2820535 on Build GRCh38 | Gain+Loss |
|
| nsv471073 | Chr.16:2687070 - 2773569 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map