Assay Details
Target Gene Details
Entrez Gene ID: | 79650 |
Gene Name: | U6 snRNA biogenesis phosphodiesterase 1 |
Gene Aliases: |
C16orf57, HVSL1, Mpn1, PN, hUsb1 |
Location: |
Chr.16:57999600-58021623 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USB1 | NM_001195302.1 | 6 | 2133 | NP_001182231.1 |
| NM_024598.3 | 7 | 2187 | NP_078874.2 | |
| XM_005256144.3 | 7 | 2062 | XP_005256201.1 | |
| XM_011523328.2 | 7 | 2163 | XP_011521630.1 | |
| XM_011523329.2 | 7 | 2045 | XP_011521631.1 | |
| AK023216.1 | 7 | 2159 | BAB14469.1 | |
| AK126223.1 | 3 | 3794 | ||
| BC004415.1 | 7 | 2123 | AAH04415.1 | |
| BC006291.2 | 8 | 2055 | AAH06291.1 | |
| BC007774.2 | 7 | 2117 | AAH07774.1 | |
| BC021554.1 | 7 | 2107 | AAH21554.1 | |
| DB364698.1 | 1 | 101 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063386 | Chr.16:57958821 - 58040306 on Build GRCh38 | Gain |
|
| esv3638716 | Chr.16:58011317 - 58046443 on Build GRCh38 | Loss |
|
| nsv572759 | Chr.16:57972385 - 58040306 on Build GRCh38 | Loss |
|
| dgv525e214 | Chr.16:58007762 - 58046443 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map