Assay Details
Target Gene Details
Entrez Gene ID: | 6430 |
Gene Name: | serine and arginine rich splicing factor 5 |
Gene Aliases: |
HRS, SFRS5, SRP40 |
Location: |
Chr.14:69767087-69772005 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRSF5 | NM_001039465.1 | NP_001034554.1 | ||
| NM_001320214.1 | NP_001307143.1 | |||
| NM_006925.4 | NP_008856.2 | |||
| XM_005267999.1 | XP_005268056.1 | |||
| XM_005268000.1 | XP_005268057.1 | |||
| XM_011537077.2 | XP_011535379.1 | |||
| XM_017021593.1 | XP_016877082.1 | |||
| AB209804.1 | BAD93041.1 | |||
| AB451250.1 | ||||
| AB451374.1 | ||||
| AK296113.1 | ||||
| AK300560.1 | ||||
| AK311805.1 | ||||
| BC018823.2 | AAH18823.1 | |||
| BC040209.1 | AAH40209.1 | |||
| BT007089.1 | AAP35752.1 | |||
| BU928782.1 | ||||
| BX247988.1 | CAD62322.1 | |||
| BX407495.2 | ||||
| BX640605.1 | CAE45711.1 | |||
| CR542167.1 | CAG46964.1 | |||
| CR542182.1 | CAG46979.1 | |||
| DC307911.1 | ||||
| DR761538.1 | ||||
| U30826.1 | AAA93070.1 | |||
| U30827.1 | AAB60405.1 | |||
| U30884.1 | AAA93074.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv456336 | Chr.14:69615239 - 70107432 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map