Assay Details
Target Gene Details
Entrez Gene ID: | 8382 |
Gene Name: | NME/NM23 family member 5 |
Gene Aliases: |
NM23-H5, NM23H5, RSPH23 |
Location: |
Chr.5:138115172-138139443 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NME5 | NM_003551.2 | NP_003542.1 | ||
| XM_005272099.1 | XP_005272156.1 | |||
| XM_006714716.3 | XP_006714779.1 | |||
| XM_017009945.1 | XP_016865434.1 | |||
| AF067724.1 | AAC64358.1 | |||
| AK312181.1 | ||||
| BC026182.2 | AAH26182.1 | |||
| BX490554.1 | 2 | 177 | ||
| KU178416.1 | ||||
| KU178417.1 | ||||
| U90450.1 | AAC69440.1 | |||
| Y14992.1 | CAA75226.1 |
More Information
Set Membership: |
|

Genomic Map