Assay Details
Target Gene Details
Entrez Gene ID: | 283316 |
Gene Name: | CD163 molecule like 1 |
Gene Aliases: |
CD163B, M160, SCARI2 |
Location: |
Chr.12:7320510-7444185 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD163L1 | NM_001297650.1 | 19 | 4441 | NP_001284579.1 |
| NM_174941.5 | 19 | 4411 | NP_777601.2 | |
| XM_011520616.1 | 18 | 4283 | XP_011518918.1 | |
| XM_011520617.2 | 18 | 4388 | XP_011518919.1 | |
| XM_011520618.2 | 17 | 3707 | XP_011518920.1 | |
| XM_011520619.1 | 18 | 3899 | XP_011518921.1 | |
| XM_011520620.1 | 17 | 3853 | XP_011518922.1 | |
| XM_011520621.2 | 12 | 2392 | XP_011518923.1 | |
| AF264014.1 | 19 | 4378 | AAF91396.1 | |
| AK058038.1 | 11 | 2048 | ||
| AK303372.1 | 19 | 4402 | ||
| BC104962.1 | 19 | 4383 | AAI04963.1 | |
| BC104964.1 | 19 | 4383 | AAI04965.1 | |
| BQ024006.1 | 2 | 220 | ||
| GQ397482.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv594 | Chr.12:7329134 - 7363501 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map