| esv2759148 | 
						
							
								Chr.3:50161045 - 50333946
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 GNAI2
							
								 LSMEM2
							
								 HYAL3
							
								 SEMA3F
							
								 HYAL2
							
								 SEMA3B
							
								 MIR566
							
								 MIR6872
							
								 IFRD2
							
								 SLC38A3
							
								 TUSC2
							
								 NAT6
							
								 RASSF1
							
								 MIR5787
							
								 SEMA3B-AS1
							
								 GNAT1
							
								 HYAL1
							
						 | 
					
				
		    		
						| nsv523560 | 
						
							
								Chr.3:50134964 - 50314769
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 GNAI2
							
								 LSMEM2
							
								 LOC107986035
							
								 SEMA3F-AS1
							
								 HYAL3
							
								 SEMA3F
							
								 SEMA3B
							
								 MIR566
							
								 MIR6872
							
								 IFRD2
							
								 SLC38A3
							
								 NAT6
							
								 MIR5787
							
								 SEMA3B-AS1
							
								 GNAT1
							
								 HYAL1
							
						 | 
					
				
		    		
						| dgv602n27 | 
						
							
								Chr.3:50140578 - 50287241
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 MIR6872
							
								 GNAI2
							
								 LSMEM2
							
								 SLC38A3
							
								 LOC107986035
							
								 SEMA3F-AS1
							
								 MIR5787
							
								 SEMA3F
							
								 SEMA3B-AS1
							
								 SEMA3B
							
								 GNAT1
							
								 MIR566
							
						 | 
					
				
		    		
						| dgv8317n54 | 
						
							
								Chr.3:50206077 - 50383123
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 GNAI2
							
								 LSMEM2
							
								 RASSF1-AS1
							
								 NPRL2
							
								 TMEM115
							
								 HYAL3
							
								 HYAL2
							
								 SEMA3B
							
								 MIR6872
							
								 IFRD2
							
								 SLC38A3
							
								 TUSC2
							
								 CACNA2D2
							
								 NAT6
							
								 RASSF1
							
								 MIR5787
							
								 ZMYND10
							
								 SEMA3B-AS1
							
								 CYB561D2
							
								 HYAL1
							
						 | 
					
				
		    		
						| nsv834693 | 
						
							
								Chr.3:50251400 - 50470533
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 GNAI2
							
								 LSMEM2
							
								 RASSF1-AS1
							
								 NPRL2
							
								 TMEM115
							
								 HYAL3
							
								 HYAL2
							
								 SEMA3B
							
								 MIR6872
							
								 IFRD2
							
								 TUSC2
							
								 CACNA2D2
							
								 NAT6
							
								 RASSF1
							
								 ZMYND10
							
								 SEMA3B-AS1
							
								 CYB561D2
							
								 HYAL1
							
						 | 
					
				
		    		
						| nsv834692 | 
						
							
								Chr.3:50161052 - 50356033
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 GNAI2
							
								 LSMEM2
							
								 RASSF1-AS1
							
								 NPRL2
							
								 TMEM115
							
								 HYAL3
							
								 SEMA3F
							
								 HYAL2
							
								 SEMA3B
							
								 MIR566
							
								 MIR6872
							
								 IFRD2
							
								 SLC38A3
							
								 TUSC2
							
								 NAT6
							
								 RASSF1
							
								 MIR5787
							
								 ZMYND10
							
								 SEMA3B-AS1
							
								 CYB561D2
							
								 GNAT1
							
								 HYAL1
							
						 | 
					
				
		    		
						| dgv8316n54 | 
						
							
								Chr.3:50115923 - 50299991
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 GNAI2
							
								 LSMEM2
							
								 RBM5
							
								 LOC107986035
							
								 SEMA3F-AS1
							
								 HYAL3
							
								 SEMA3F
							
								 SEMA3B
							
								 MIR566
							
								 MIR6872
							
								 IFRD2
							
								 SLC38A3
							
								 NAT6
							
								 MIR5787
							
								 SEMA3B-AS1
							
								 GNAT1
							
								 HYAL1
							
						 | 
					
				
		    		
						| nsv470561 | 
						
							
								Chr.3:50121341 - 50547568
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 LSMEM2
							
								 RASSF1-AS1
							
								 NPRL2
							
								 LOC107986035
							
								 HYAL3
							
								 SEMA3F
							
								 HYAL2
							
								 SEMA3B
							
								 MIR566
							
								 IFRD2
							
								 SLC38A3
							
								 NAT6
							
								 ZMYND10
							
								 GNAT1
							
								 GNAI2
							
								 SEMA3F-AS1
							
								 TMEM115
							
								 MIR6872
							
								 TUSC2
							
								 CACNA2D2
							
								 RASSF1
							
								 MIR5787
							
								 SEMA3B-AS1
							
								 CYB561D2
							
								 HYAL1
							
						 | 
					
				
		    		
						| nsv523662 | 
						
							
								Chr.3:50107518 - 50547568
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 LSMEM2
							
								 RASSF1-AS1
							
								 RBM5
							
								 NPRL2
							
								 LOC107986035
							
								 HYAL3
							
								 SEMA3F
							
								 HYAL2
							
								 SEMA3B
							
								 MIR566
							
								 IFRD2
							
								 SLC38A3
							
								 NAT6
							
								 ZMYND10
							
								 GNAT1
							
								 GNAI2
							
								 SEMA3F-AS1
							
								 TMEM115
							
								 MIR6872
							
								 TUSC2
							
								 CACNA2D2
							
								 RASSF1
							
								 MIR5787
							
								 SEMA3B-AS1
							
								 CYB561D2
							
								 HYAL1
							
						 | 
					
				
		    		
						| nsv590255 | 
						
							
								Chr.3:50210391 - 50399396
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 GNAI2
							
								 LSMEM2
							
								 RASSF1-AS1
							
								 NPRL2
							
								 TMEM115
							
								 HYAL3
							
								 HYAL2
							
								 SEMA3B
							
								 MIR6872
							
								 IFRD2
							
								 SLC38A3
							
								 TUSC2
							
								 CACNA2D2
							
								 NAT6
							
								 RASSF1
							
								 MIR5787
							
								 ZMYND10
							
								 SEMA3B-AS1
							
								 CYB561D2
							
								 HYAL1
							
						 | 
					
				
		    		
						| nsv954855 | 
						
							
								Chr.3:50152468 - 50439169
							    on Build GRCh38
						
							
						 | 
						Deletion | 
						
							
								 LSMEM2
							
								 RASSF1-AS1
							
								 NPRL2
							
								 LOC107986035
							
								 HYAL3
							
								 SEMA3F
							
								 HYAL2
							
								 SEMA3B
							
								 MIR566
							
								 IFRD2
							
								 SLC38A3
							
								 NAT6
							
								 ZMYND10
							
								 GNAT1
							
								 GNAI2
							
								 SEMA3F-AS1
							
								 TMEM115
							
								 MIR6872
							
								 TUSC2
							
								 CACNA2D2
							
								 RASSF1
							
								 MIR5787
							
								 SEMA3B-AS1
							
								 CYB561D2
							
								 HYAL1
							
						 |