Assay Details
Target Gene Details
Entrez Gene ID: | 55335 |
Gene Name: | nipsnap homolog 3B |
Gene Aliases: |
FP944, NIPSNAP3, SNAP1 |
Location: |
Chr.9:104763741-104777764 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NIPSNAP3B | NM_018376.3 | 6 | 2193 | NP_060846.2 |
| NR_130759.1 | 5 | 1982 | ||
| NR_130760.1 | ||||
| XM_011518839.1 | 5 | 2043 | XP_011517141.1 | |
| XM_017014889.1 | XP_016870378.1 | |||
| BC017914.1 | AAH17914.1 | |||
| BX107655.1 | ||||
| BX647652.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891706 | Chr.9:104480016 - 105304573 on Build GRCh38 | Gain |
|
| esv3621343 | Chr.9:104761673 - 104776613 on Build GRCh38 | Loss |
|
| esv3891705 | Chr.9:104391828 - 106660921 on Build GRCh38 | Loss |
|

Genomic Map