Assay Details
Target Gene Details
Entrez Gene ID: | 7375 |
Gene Name: | ubiquitin specific peptidase 4 |
Gene Aliases: |
UNP, Unph |
Location: |
Chr.3:49277144-49340103 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP4 | NM_003363.3 | 22 | 3278 | NP_003354.2 |
| NM_199443.2 | 21 | 3137 | NP_955475.1 | |
| AI887410.1 | 1 | 156 | ||
| AK222725.1 | 21 | 3067 | BAD96445.1 | |
| BU632996.1 | 1 | 441 | ||
| DB534804.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008750 | Chr.3:49171382 - 49324968 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map