Assay Details
Target Gene Details
Entrez Gene ID: | 435 |
Gene Name: | argininosuccinate lyase |
Gene Aliases: |
ASAL |
Location: |
Chr.7:66075789-66093343 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASL | NM_000048.3 | NP_000039.2 | ||
| NM_001024943.1 | 1 | 199 | NP_001020114.1 | |
| NM_001024944.1 | 1 | 199 | NP_001020115.1 | |
| NM_001024946.1 | 1 | 199 | NP_001020117.1 | |
| AK300519.1 | ||||
| AK310912.1 | 1 | 205 | ||
| AK316334.1 | 1 | 95 | ||
| BC008195.1 | AAH08195.1 | |||
| BC033146.1 | AAH33146.1 | |||
| BQ949510.1 | 1 | 199 | ||
| BX328836.2 | ||||
| J03058.1 | AAA51787.1 | |||
| M14218.1 | 1 | 79 | AAA51786.1 | |
| M57638.1 | AAA51788.1 | |||
| Y00753.1 | CAA68722.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761079 | Chr.7:65601260 - 66224746 on Build GRCh38 | Gain+Loss |
|
| esv2664145 | Chr.7:65814760 - 66395282 on Build GRCh38 | Deletion |
|
| nsv1021186 | Chr.7:66027614 - 66198451 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map