Assay Details
Target Gene Details
Entrez Gene ID: | 92211 |
Gene Name: | cadherin related family member 1 |
Gene Aliases: |
CORD15, PCDH21, PRCAD, RP65 |
Location: |
Chr.10:84194635-84219621 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDHR1 | NM_001171971.2 | NP_001165442.1 | ||
| NM_033100.3 | NP_149091.1 | |||
| XM_011540337.1 | XP_011538639.1 | |||
| XM_011540338.1 | XP_011538640.1 | |||
| XM_011540339.1 | XP_011538641.1 | |||
| XM_011540340.2 | XP_011538642.1 | |||
| AB053448.1 | BAB61905.1 | |||
| AB593154.1 | ||||
| AB593155.1 | ||||
| AK124752.1 | ||||
| BC038799.1 | AAH38799.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046749 | Chr.10:83270389 - 84629720 on Build GRCh38 | Gain |
|
| nsv551753 | Chr.10:84187538 - 84215514 on Build GRCh38 | Loss |
|
| nsv551745 | Chr.10:83260366 - 84632635 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map