Assay Details
Target Gene Details
Entrez Gene ID: | 23154 |
Gene Name: | neurochondrin |
Gene Aliases: |
- |
Location: |
Chr.1:35557792-35566779 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCDN | NM_001014839.1 | NP_001014839.1 | ||
| NM_001014841.1 | NP_001014841.1 | |||
| NM_014284.2 | NP_055099.1 | |||
| AB011179.1 | ||||
| AB018739.2 | BAA85384.2 | |||
| AB018740.2 | BAA85385.2 | |||
| AL536768.3 | ||||
| BC024592.1 | AAH24592.1 | |||
| BI545092.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012556 | Chr.1:35413704 - 35703660 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map