Assay Details
Target Gene Details
Entrez Gene ID: | 3073 |
Gene Name: | hexosaminidase subunit alpha |
Gene Aliases: |
TSD |
Location: |
Chr.15:72343435-72376473 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEXA | NM_000520.5 | 14 | 2384 | NP_000511.2 |
| NM_001318825.1 | 14 | 2417 | NP_001305754.1 | |
| BM668409.1 | 1 | 368 | ||
| M13520.1 | 14 | 1767 | AAA51827.1 |
More Information
Set Membership: |
|

Genomic Map