Assay Details
Target Gene Details
Entrez Gene ID: | 1831 |
Gene Name: | TSC22 domain family member 3 |
Gene Aliases: |
DIP, DSIPI, GILZ, TSC-22R |
Location: |
Chr.X:107713221-107777329 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSC22D3 | NM_001015881.1 | 3 | 454 | NP_001015881.1 |
| NM_001318468.1 | 4 | 922 | NP_001305397.1 | |
| NM_001318470.1 | 4 | 908 | NP_001305399.1 | |
| NM_004089.3 | 3 | 659 | NP_004080.2 | |
| NM_198057.2 | 3 | 975 | NP_932174.1 | |
| XM_005262099.1 | 5 | 936 | XP_005262156.1 | |
| XM_005262100.1 | 4 | 811 | XP_005262157.1 | |
| XM_005262102.1 | 4 | 804 | XP_005262159.1 | |
| XM_005262103.3 | 4 | 914 | XP_005262160.1 | |
| XM_011530884.1 | 4 | 1010 | XP_011529186.1 | |
| XM_017029335.1 | 4 | 746 | XP_016884824.1 | |
| AB025432.1 | 3 | 642 | BAB18680.1 | |
| AF153603.1 | 3 | 637 | AAD41085.1 | |
| AF183393.1 | 3 | 645 | AAD56234.1 | |
| AF228339.1 | 3 | 649 | AAG12456.1 | |
| AK092645.1 | 4 | 861 | BAC03934.1 | |
| AK092669.1 | 3 | 947 | ||
| AK127938.1 | 3 | 599 | ||
| AK300535.1 | 4 | 786 | ||
| AK311834.1 | ||||
| AL110191.1 | 3 | 658 | CAB53669.1 | |
| AY007119.1 | 3 | 558 | ||
| BC018148.1 | 3 | 341 | ||
| BC072446.1 | 3 | 653 | AAH72446.1 | |
| BX647854.1 | 4 | 908 | ||
| CR533450.1 | 3 | 397 | CAG38481.1 | |
| CR933650.1 | 3 | 661 | CAI45951.1 | |
| Z50781.1 | CAA90644.1 |
More Information
Set Membership: |
|

Genomic Map