Assay Details
Target Gene Details
Entrez Gene ID: | 6518 |
Gene Name: | solute carrier family 2 member 5 |
Gene Aliases: |
GLUT-5, GLUT5 |
Location: |
Chr.1:9036941-9093407 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC2A5 | NM_003039.2 | NP_003030.1 | ||
| XM_005263491.3 | XP_005263548.1 | |||
| XM_017002133.1 | XP_016857622.1 | |||
| XM_017002134.1 | XP_016857623.1 | |||
| XM_017002135.1 | XP_016857624.1 | |||
| XM_017002136.1 | XP_016857625.1 | |||
| XM_017002137.1 | XP_016857626.1 | |||
| XM_017002138.1 | XP_016857627.1 | |||
| XM_017002139.1 | XP_016857628.1 | |||
| XM_017002140.1 | XP_016857629.1 | |||
| XM_017002141.1 | XP_016857630.1 | |||
| XM_017002142.1 | XP_016857631.1 | |||
| AK290398.1 | ||||
| AK294368.1 | ||||
| AK300091.1 | ||||
| AK300477.1 | ||||
| AK302047.1 | ||||
| AK302057.1 | ||||
| AK302137.1 | ||||
| BC001692.2 | AAH01692.1 | |||
| BC001820.2 | AAH01820.1 | |||
| M55531.1 | AAA52570.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004025 | Chr.1:9000983 - 9069086 on Build GRCh38 | Gain |
|
| nsv545347 | Chr.1:9020352 - 9072997 on Build GRCh38 | Loss |
|
| nsv831981 | Chr.1:9031658 - 9187508 on Build GRCh38 | Loss |
|
| dgv15n100 | Chr.1:8992095 - 9056684 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map