Assay Details
Target Gene Details
Entrez Gene ID: | 2834 |
Gene Name: | prolactin releasing hormone receptor |
Gene Aliases: |
GPR10, GR3, PrRPR |
Location: |
Chr.10:118593404-118595648 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRLHR | NM_004248.2 | 2 | 460 | NP_004239.1 |
| AB015745.1 | 1 | 321 | BAA31159.1 | |
| AK290461.1 | 2 | 460 | ||
| AK313269.1 | 2 | 462 | ||
| BC095539.1 | 1 | 446 | AAH95539.1 | |
| BC101490.1 | 1 | 395 | AAI01491.1 | |
| BC101492.1 | 1 | 395 | AAI01493.1 | |
| DA262254.1 | 2 | 486 | ||
| EF577402.1 | 1 | 321 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832006 | Chr.10:118557873 - 118759411 on Build GRCh38 | Gain |
|
| nsv7569 | Chr.10:118578807 - 118611109 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map