Assay Details
Target Gene Details
Entrez Gene ID: | 10849 |
Gene Name: | CD3e molecule associated protein |
Gene Aliases: |
ASE-1, ASE1, CAST, PAF49 |
Location: |
Chr.19:45406129-45410766 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD3EAP | NM_001297590.1 | 3 | 2042 | NP_001284519.1 |
| NM_012099.1 | 3 | 2036 | NP_036231.1 | |
| XM_017026180.1 | 3 | 2137 | XP_016881669.1 | |
| XM_017026181.1 | 3 | 2207 | XP_016881670.1 | |
| AF017633.1 | 3 | 1560 | AAD41158.1 | |
| AK292183.1 | 3 | 1601 | ||
| BC038992.1 | 3 | 1564 | AAH38992.1 | |
| BC108889.2 | 3 | 1801 | AAI08890.1 | |
| U86751.1 | 3 | 2036 | AAB68608.1 |
Target Gene Details
Entrez Gene ID: | 2067 |
Gene Name: | ERCC excision repair 1, endonuclease non-catalytic subunit |
Gene Aliases: |
COFS4, RAD10, UV20 |
Location: |
Chr.19:45407333-45478866 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERCC1 | NM_001166049.1 | 9 | 1127 | NP_001159521.1 |
| NM_001983.3 | 10 | 1199 | NP_001974.1 | |
| XM_017026460.1 | 10 | 1600 | XP_016881949.1 | |
| XM_017026461.1 | 11 | 1229 | XP_016881950.1 | |
| XM_017026462.1 | 11 | 1194 | XP_016881951.1 | |
| XM_017026463.1 | 10 | 1186 | XP_016881952.1 | |
| XM_017026464.1 | 10 | 1187 | XP_016881953.1 | |
| XM_017026465.1 | 9 | 1528 | XP_016881954.1 | |
| XM_017026466.1 | 10 | 1162 | XP_016881955.1 | |
| AK092039.1 | 9 | 937 | ||
| BM011724.1 | 2 | 254 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833845 | Chr.19:45308814 - 45457371 on Build GRCh38 | Loss |
|
| nsv833846 | Chr.19:45381005 - 45469089 on Build GRCh38 | Loss |
|
| nsv428365 | Chr.19:45358027 - 45469089 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map