Assay Details
Target Gene Details
Entrez Gene ID: | 8986 |
Gene Name: | ribosomal protein S6 kinase A4 |
Gene Aliases: |
MSK2, RSK-B, S6K-alpha-4 |
Location: |
Chr.11:64359153-64372215 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS6KA4 | NM_001006944.1 | NP_001006945.1 | ||
| NM_001300802.1 | NP_001287731.1 | |||
| NM_001318361.1 | NP_001305290.1 | |||
| NM_003942.2 | NP_003933.1 | |||
| XM_005274380.1 | XP_005274437.1 | |||
| AF074715.1 | AAC67395.1 | |||
| AJ010119.1 | CAA09009.1 | |||
| AK223561.1 | BAD97281.1 | |||
| AK292163.1 | ||||
| AK298507.1 | ||||
| AK313567.1 | ||||
| BC028079.1 | ||||
| BC047896.1 | AAH47896.1 | |||
| BI253655.1 | ||||
| BI523588.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832187 | Chr.11:64354497 - 64544828 on Build GRCh38 | Loss |
|
| nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss |
|
| dgv13n64 | Chr.11:64329761 - 64362250 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map