Assay Details
Target Gene Details
Entrez Gene ID: | 8543 |
Gene Name: | LIM domain only 4 |
Gene Aliases: |
- |
Location: |
Chr.1:87328468-87348923 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMO4 | NM_006769.3 | 1 | 533 | NP_006760.1 |
| XM_005271291.3 | 1 | 133 | XP_005271348.1 | |
| AI962216.1 | 1 | 81 | ||
| AK292578.1 | 1 | 135 | ||
| AW162477.1 | 1 | 77 | ||
| BC017673.1 | AAH17673.1 | |||
| BU580860.1 | ||||
| U24576.1 | 1 | 533 | AAB51073.1 |
More Information
Set Membership: |
|

Genomic Map